Learn More
Cole-Parmer Pressure Regulator, 1/4" NPT(F), 2-120 psi
Catalog No. NC0749549 Shop All Cole-Parmer Products
Encompass_Preferred
Marsh Bellofram 960-015-000 Pressure Regulator, 1/4" NPT(F), 2-120 psi, Provide precision control in adverse conditionsUse with clean, dry airIdeal for gas mixing, valve operations, and calibration stands
Catalog No. NC0749549 Supplier Cole-Parmer Supplier No. 6882510
Marsh Bellofram 960-015-000 Pressure Regulator, 1/4" NPT(F), 2-120 psi, Provide precision control in adverse conditionsUse with clean, dry airIdeal for gas mixing, valve operations, and calibration stands