Learn More
SPECIALIZEDRX MARSHMALLOW NAT SD POWDER 100G
Catalog No. NC3863036
Encompass
A natural/ spray-dried/ anhydrous, water soluble/miscible powder flavor concentrate.Contains no allergensDye-free and gluten-freeEasily incorporates into hard lozenge/candy bases/ troche bases/ rapid dissolve tablets or oral suspensions.Free-flowingRelatively heat tolerantSpray-driedSuggested concentration range: 1-2 percentTaste and aromaVegan compliant /contains no animal products/
Catalog No. NC3863036 Supplier SPECIALIZEDRX Supplier No. F017340
A natural/ spray-dried/ anhydrous, water soluble/miscible powder flavor concentrate.Contains no allergensDye-free and gluten-freeEasily incorporates into hard lozenge/candy bases/ troche bases/ rapid dissolve tablets or oral suspensions.Free-flowingRelatively heat tolerantSpray-driedSuggested concentration range: 1-2 percentTaste and aromaVegan compliant /contains no animal products/