Learn More
SPECIALIZEDRX MARSHMALLOW WS NAT LIQUID30ML
Catalog No. NC3863032
Encompass
A natural / anhydrous / water soluble/miscible liquid flavor concentrate.Contains no allergensGluten-freeSuggested concentration range: 0.1-0.5 percentTaste and aromaVegan compliant /contains no animal products/24-month shelf life from date of manufacture when stored in original, unopened container in a cool dry place
Catalog No. NC3863032 Supplier SPECIALIZEDRX Supplier No. F011898
A natural / anhydrous / water soluble/miscible liquid flavor concentrate.Contains no allergensGluten-freeSuggested concentration range: 0.1-0.5 percentTaste and aromaVegan compliant /contains no animal products/24-month shelf life from date of manufacture when stored in original, unopened container in a cool dry place