Learn More
Diagnostic Biosystems MART-1/MELAN A MART-1/Melan-A, Melanoma Marker
Catalog No. 502407609
Encompass_Preferred
This antibody is specific to a protein of 20 kDa known as MART-1 (melanoma antigen recognized by T cells-1) or Melan-A. This antibody does not crossreact with MAGE-1or tyrosinase. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. This antibody stains melanomas and other tumors showing melanocytic differentiation.
Catalog No. 50-240-7609 Supplier Diagnostic Biosystems Supplier No. PDM153
This antibody is specific to a protein of 20 kDa known as MART-1 (melanoma antigen recognized by T cells-1) or Melan-A. This antibody does not crossreact with MAGE-1or tyrosinase. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. This antibody stains melanomas and other tumors showing melanocytic differentiation.