Learn More
Charles River Labs Cell Solutions MART-1 peptide | 1 mg/vial
Supplier:  Charles River Labs Cell Solutions ANTIGEN1109
Encompass
MART-1 ia a tumor associated antigen found in melanoma. This peptide binds to HLA-A*02:01 and has the sequence ELAGIGILTV. T cells specific for this peptide can be found in our listing of antigen specific T cells.
Catalog No. NC1711920