Learn More
Enzo Life Sciences MART-1 Recombinant monoclonal antibody (rMLANA/788) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3140100
Encompass_Preferred
Clone: rMLANA/788. Isotype: lgG1κ. Immunogen: Recombinant full length human MART-1. Species reactivity: Human, Mouse, Rat. Applications: Flow Cytometry, IF, IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA (US sourced) and 0.05% sodium azide.
Catalog No. 50-201-2847