Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2315MSM8P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: MLANA/788. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: SK-MEL-13, SK-MEL-19 Melanoma cell lines; Melanomas. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human MLANA protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 20-22kDa (doublet).Chromosome: 9p24.1. Gene Symbol: MLANA. Gene ID: 2315. Ensemble: ENSG00000120215. Uniprot: NP_005502.
Catalog No. 50-188-4693