Learn More
Medchemexpress LLC Martinostat hydrochloride | 1629052-58-9 | 99.0% | 390.95 | C22H31ClN2O2 | 10 MG
SDP
Catalog No. 5000285421
Encompass_Preferred
Martinostat hydrochloride is a histone deacetylase (HDAC) inhibitor that can be labeled with radionuclides for quantitative in vivo imaging of HDACs in the central nervous system and major peripheral organs.
  • Histone deacetylase inhibitor suitable for imaging applications.
  • Can be labeled with radionuclides for quantitative in vivo imaging.
  • High purity (99.01%).
  • Available as solid (5-500 mg; larger sizes by quote) and as 10 mM solution in DMSO.
  • Storage: 4°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-85421 Supplier Medchemexpress LLC Supplier No. HY160092A10MG
Martinostat hydrochloride is a histone deacetylase (HDAC) inhibitor that can be labeled with radionuclides for quantitative in vivo imaging of HDACs in the central nervous system and major peripheral organs.
  • Histone deacetylase inhibitor suitable for imaging applications.
  • Can be labeled with radionuclides for quantitative in vivo imaging.
  • High purity (99.01%).
  • Available as solid (5-500 mg; larger sizes by quote) and as 10 mM solution in DMSO.
  • Storage: 4°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).