Learn More
VITRO VIVO BIOTECH LLC VitroView™ Martius Scarlet Blue (MSB) Stain Kit
Supplier:  VITRO VIVO BIOTECH LLC VB3036
Encompass_Preferred
The Martius Scarlet Blue (MSB) technique proves to be a dependable approach for distinguishing between fresh and aged fibrin clusters. It achieves this differentiation by employing Martius Yellow, which selectively stains erythrocytes and early fibrin deposits. The staining process involves the utilization of dyes of varying molecular sizes: Crystal Scarlet, a medium-sized molecule dye, and larger molecule dyes like Phosphotungstic Acid and Aniline Blue. This trichrome-type staining results in red-colored fibrin and muscle tissues. On the other hand, collagen and older fibrin clusters take on a blue hue.The Martius Scarlet Blue (MSB) staining method serves as a valuable tool for visualizing fibrin, particularly the older clusters. The associated stain kit is designed to facilitate the histological demonstration of fibrin, muscle, collagen, and erythrocytes.
Catalog No. NC2891501