Learn More
Martor Secunorm Smartcut Mdp
Catalog No. NC1054225
Encompass_Preferred
The Smartcut is a disposable, semi automatic retractable spring loaded knife for cutting general industrial materials. This version has a stainless blade installed and is made of metal infused plastic.-Semi automatic retractable blade for safety-Ergonomically friendly ambidextrous slider mechanism to engage the blade-Durable body made of metal infused plastic-Safer alternative to fixed blade utility knives for general cutting tasks
Catalog No. NC1054225 Supplier Martor Supplier No. 110700.02
The Smartcut is a disposable, semi automatic retractable spring loaded knife for cutting general industrial materials. This version has a stainless blade installed and is made of metal infused plastic.-Semi automatic retractable blade for safety-Ergonomically friendly ambidextrous slider mechanism to engage the blade-Durable body made of metal infused plastic-Safer alternative to fixed blade utility knives for general cutting tasks