Learn More
Aviva Systems Biology Corp MARVELD2 Antibody - middle region (ARP42885_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42885_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MARVELD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MARVELD2. Alternate names: DFNB49, FLJ30532, MARVD2, MRVLDC2, TricApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58351