Learn More
Advanced Materials Technology MARVELXACT PEEKSIL 75 ID X
Supplier:  Advanced Materials Technology PS7075350
Encompass_Preferred
Advanced Materials Technology’s MarvelXACT finger-tight UHPLC connectors with volume definitions easily identifies dispersion volume contribution. Haptic “click” indicates optimum torque and zero dead volume connection. Connector and tubing for both pre and post column installation
Catalog No. 50-255-5558