Learn More
Advanced Materials Technology MARVELXACT PEEKSIL 75 ID X
Catalog No. 502555557
Encompass_Preferred
Advanced Materials Technology’s MarvelXACT finger-tight UHPLC connectors with volume definitions easily identifies dispersion volume contribution. Haptic “click” indicates optimum torque and zero dead volume connection. Connector and tubing for both pre and post column installation
Catalog No. 50-255-5557 Supplier Advanced Materials Technology Supplier No. PS7075600
Advanced Materials Technology’s MarvelXACT finger-tight UHPLC connectors with volume definitions easily identifies dispersion volume contribution. Haptic “click” indicates optimum torque and zero dead volume connection. Connector and tubing for both pre and post column installation