Learn More
Advanced Materials Technology MARVELXACT PLS 50 X 350MM
Catalog No. 502555573
Encompass_Preferred
Advanced Materials Technology’s MarvelXACT finger-tight UHPLC connectors with volume definitions easily identifies dispersion volume contribution. Haptic “click” indicates optimum torque and zero dead volume connection. Connector and tubing for both pre and post column installation
Catalog No. 50-255-5573 Supplier Advanced Materials Technology Supplier No. PL7050350
Advanced Materials Technology’s MarvelXACT finger-tight UHPLC connectors with volume definitions easily identifies dispersion volume contribution. Haptic “click” indicates optimum torque and zero dead volume connection. Connector and tubing for both pre and post column installation