Learn More
IDEX Health & Science Upchurch MarvelXACT™ Stainless Steel 100µm ID x 500mm Length
Catalog No. NC3432488
Encompass_Preferred
10-32 threaded torque limiting fittingsMarvelXACT™ Connection Systems have been expertly designed to eliminate the risk of under or over-tightening with our patented torque limiting mechanism. This unique feature emits a haptic click feedback when it reaches the optimum torque.
Catalog No. NC3432488 Supplier IDEX Health & Science Upchurch Supplier No. UPFS7100500
10-32 threaded torque limiting fittingsMarvelXACT™ Connection Systems have been expertly designed to eliminate the risk of under or over-tightening with our patented torque limiting mechanism. This unique feature emits a haptic click feedback when it reaches the optimum torque.