Learn More
Shimadzu Scientific Instruments MarvelXACT Stainless Steel 125um ID X 600mm Tubing w/Fittings
Catalog No. NC3313568
Encompass
MarvelXACT is comprised of flexible 1/32" OD stainless steel tubing for easy routing through the LC system and can be connected and disconnected multiple times. The 1/16" tubing ends and integrated fittings are compatible with any 10-32 coned port, and are finger-tight — no tool required for UHPLC connections up to 19,000 psi. The fitting produces a haptic "click" feedback when it reaches the optimum torque to eliminate the risk of under- or over-tightening. The PEEK tubing end seals at the port bottom which eliminates dead volume and minimizes carryover risk, peak tailing, and peak broadening. Tubing volume: 7.363uL.
Catalog No. NC3313568 Supplier Shimadzu Scientific Instruments Supplier No. 2209124610
MarvelXACT is comprised of flexible 1/32" OD stainless steel tubing for easy routing through the LC system and can be connected and disconnected multiple times. The 1/16" tubing ends and integrated fittings are compatible with any 10-32 coned port, and are finger-tight — no tool required for UHPLC connections up to 19,000 psi. The fitting produces a haptic "click" feedback when it reaches the optimum torque to eliminate the risk of under- or over-tightening. The PEEK tubing end seals at the port bottom which eliminates dead volume and minimizes carryover risk, peak tailing, and peak broadening. Tubing volume: 7.363uL.