Learn More
Newcomer Supply MAS Adhesive Slides, 100 slides/box, Extremely strong adhesion with a hydrophilic surface
SDP
Catalog No. NC1693867
Encompass
The MAS slide has remarkably improved adhesion to tissue sections and cells due to the amino groups that bind to the tissue section have a much higher density on the slide than could be achieved by either poly-L-lysine or silane coatings. This adhesive coated slide is excellent for the most challenging tissues and applications: demading IHC with antigen retrieval, veterinary animal tissue, difficult tissue (dry, fatty or hard tissue such as bones, nails, breast, eyes and brain). Size: 75x25, 1mm thick; Surface Wettability: Hydrophilic; Edge Treatment: 90° Corner; Unit: 100 slides/box, 10 boxes/case; Color: White.
This item is not returnable. View return policy
Catalog No. NC1693867 Supplier Newcomer Supply Supplier No. 5077W90BX
The MAS slide has remarkably improved adhesion to tissue sections and cells due to the amino groups that bind to the tissue section have a much higher density on the slide than could be achieved by either poly-L-lysine or silane coatings. This adhesive coated slide is excellent for the most challenging tissues and applications: demading IHC with antigen retrieval, veterinary animal tissue, difficult tissue (dry, fatty or hard tissue such as bones, nails, breast, eyes and brain). Size: 75x25, 1mm thick; Surface Wettability: Hydrophilic; Edge Treatment: 90° Corner; Unit: 100 slides/box, 10 boxes/case; Color: White.