Learn More
Thermo Scientific™ MAS™ Alcohol and Ammonia Controls, Vista Vials, 5 x 2.0mL
Monitor alcohol and/or ammonia in human serum with bi-level, assayed liquid MAS Alcohol and Ammonia Controls.
Supplier:  Thermo Scientific™ 10014443
Catalog No. 23-640-123
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Monitor alcohol and/or ammonia in human serum with bi-level, assayed liquid Thermo Scientific™ MAS™ Alcohol and Ammonia Controls. Available in a convenient Vista™ vial format, this ready-to-use product is packed in convenient multipacks.
  • Human-serum based matrix
  • Normal ammonia values in level 1
  • Assayed values available for most major analyzers
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions