Learn More
Thermo Scientific™ MAS™ Bilirubin Controls, Assayed, Level 2, 6 x 5mL
Monitor the performance of total, conjugated, unconjugated and neonatal Bilirubin with tri-level, assayed MAS Bilirubin Controls.
Supplier:  Thermo Scientific™ BC102
Catalog No. CB22002
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Monitor the performance of total, conjugated, unconjugated and neonatal Bilirubin with tri-level, assayed Thermo Scientific™ MAS™ Bilirubin Controls. These controls are also available in a convenient Vista™ vial format.
  • Liquid, ready-to-use; no additional preparation steps required
  • Bovine serum matrix
  • Assayed values available for most major analyzers
  • Elevated bilirubin in Level 3 for neonatal applications
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories