Learn More
Thermo Scientific™ MAS™ Bilirubin Controls, Assayed, Level 3, 6 x 5mL
GSA_VA
Monitor the performance of total, conjugated, unconjugated and neonatal Bilirubin with tri-level, assayed MAS Bilirubin Controls.
Supplier:  Thermo Scientific™ BC103
Catalog No. CB33003
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Monitor the performance of total, conjugated, unconjugated and neonatal Bilirubin with tri-level, assayed Thermo Scientific™ MAS™ Bilirubin Controls. These controls are also available in a convenient Vista™ vial format.
  • Liquid, ready-to-use; no additional preparation steps required
  • Bovine serum matrix
  • Assayed values available for most major analyzers
  • Elevated bilirubin in Level 3 for neonatal applications
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories