Learn More
Thermo Scientific™ MAS™ CardioImmune™ XL Controls
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls.
Supplier: Thermo Scientific™ 10014180
Description
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls. These controls are also available in a convenient Vista™ vial format.
- Human serum matrix
- Offers 4 distinctive levels
- Assayed values available for most major analyzers
- Available in a tri-level multipack