Learn More
Thermo Scientific™ MAS™ CardioImmune™ XL Controls
GSA_VA
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls.
Supplier:  Thermo Scientific™ CAIXL1
 View more versions of this product
Catalog No. 23-640-000
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls. These controls are also available in a convenient Vista™ vial format.
  • Human serum matrix
  • Offers 4 distinctive levels
  • Assayed values available for most major analyzers
  • Available in a tri-level multipack
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions