Learn More
Thermo Scientific™ MAS™ CardioImmune™ XL Controls
GSA_VA
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls.
Supplier:  Thermo Scientific™ CAIXL2
 View more versions of this product
Catalog No. 23-640-001
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls. These controls are also available in a convenient Vista™ vial format.
  • Human serum matrix
  • Offers 4 distinctive levels
  • Assayed values available for most major analyzers
  • Available in a tri-level multipack
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions