Learn More
Thermo Scientific™ MAS™ Cerebrospinal Fluid Control
Monitor the performance of quantitative and manual chemical analysis of human cerebrospinal fluid with Thermo Scientific™ MAS™ Cerebrospinal Fluid Control.
| Catalog Number | Mfr. No. | Description | dimQuantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | dimQuantity | Price | Quantity & Availability | ||||
|
23-640-122
|
Thermo Scientific™
10014444 |
N/A | N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-038-386
![]() |
Thermo Scientific™
CSFMP |
N/A | N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
Description
Monitor the performance of quantitative and manual chemical analysis of human cerebrospinal fluid with Thermo Scientific™ MAS™ Cerebrospinal Fluid Control, a bi-level control assayed for cellulose space acetate and agarose gel electrophoresis.
- Requires no additional preparation
- Eliminates variables due to reconstitution, mixing errors, or uncertain water quality
- Contains 18 analytes† including Glutamine and Potassium
- Human-serum based
- Assayed ranges for both chemistry and protein electrophoresis
- Assayed for most major analyzers
- Refer to package insert for specific analyte and stability claims