Learn More
Thermo Scientific™ MAS™ Cerebrospinal Fluid Control
GSA_VA
Monitor the performance of quantitative and manual chemical analysis of human cerebrospinal fluid with Thermo Scientific™ MAS™ Cerebrospinal Fluid Control.
Supplier:  Thermo Scientific™ CSFMP
 View more versions of this product
Catalog No. 23-038-386
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Monitor the performance of quantitative and manual chemical analysis of human cerebrospinal fluid with Thermo Scientific™ MAS™ Cerebrospinal Fluid Control, a bi-level control assayed for cellulose space acetate and agarose gel electrophoresis.
  • Requires no additional preparation
  • Eliminates variables due to reconstitution, mixing errors, or uncertain water quality
  • Contains 18 analytes† including Glutamine and Potassium
  • Human-serum based
  • Assayed ranges for both chemistry and protein electrophoresis
  • Assayed for most major analyzers
  • Refer to package insert for specific analyte and stability claims
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories