Learn More
Thermo Scientific™ MAS™ chemTRAK™ H General Chemistry Controls
Ensure accurate results in general chemistry and TDM methods with comprehensive, multi-analyte Thermo Scientific™ MAS™ chemTRAK™ H General Chemistry Controls.
Supplier: Thermo Scientific™ 10014152
Description
Ensure accurate results in general chemistry and TDM methods with comprehensive, multi-analyte Thermo Scientific™ MAS™ chemTRAK™ H General Chemistry Controls. These liquid, ready-to-use controls are available in a convenient Vista™ vial format in both assayed and unassayed formats.
- Human serum matrix
- Assayed values available for most major analyzers
- Refer to package insert for specific analyte and stability claims