Learn More
Thermo Scientific™ MAS™ chemTRAK™ H General Chemistry Controls
GSA_VA
Ensure accurate results in general chemistry and TDM methods with comprehensive, multi-analyte Thermo Scientific™ MAS™ chemTRAK™ H General Chemistry Controls.
Supplier:  Thermo Scientific™ CHA1
 View more versions of this product
Catalog No. 23-640-065
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Ensure accurate results in general chemistry and TDM methods with comprehensive, multi-analyte Thermo Scientific™ MAS™ chemTRAK™ H General Chemistry Controls. These liquid, ready-to-use controls are available in a convenient Vista™ vial format in both assayed and unassayed formats.
  • Human serum matrix
  • Assayed values available for most major analyzers
  • Refer to package insert for specific analyte and stability claims
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions