Learn More
Thermo Scientific™ MAS™ DOA TOTAL Controls
Monitor immunoassay screening procedures with multi-constituent urine toxicology controls.
| Catalog Number | Mfr. No. | dimLevel | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | dimLevel | Price | Quantity & Availability | |||||
23-139-331
![]() |
Thermo Scientific™
DOAT1 |
Pack of 6 for $598.00
|
N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-139-332
![]() |
Thermo Scientific™
DOAT2 |
Pack of 6 for $889.00
|
N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-139-333
![]() |
Thermo Scientific™
DOAT3 |
Pack of 6 for $889.00
|
N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-139-334
![]() |
Thermo Scientific™
DOAT4 |
Pack of 6 for $889.00
|
N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-139-335
![]() |
Thermo Scientific™
DOAT5 |
Pack of 6 for $889.00
|
N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-139-336
![]() |
Thermo Scientific™
DOAT6 |
Pack of 6 for $905.00
|
N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
|
23-139-330
|
Thermo Scientific™
DOATMP |
Pack of 6 for $915.00
|
N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
Description
Monitor immunoassay screening procedures with liquid, ready-to-use Thermo Scientific™ MAS™ DOA TOTAL Controls. These are multi-constituent urine toxicology controls, offering 19 analytes with four distinct levels at drug concentrations 25% below and above commonly used screening and SAMHSA cutoffs. A drug-free level and high positive level are also available (six levels total).
- Human urine based matrix
- GC/MS confirmation values available
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.