Learn More
Thermo Scientific™ MAS™ DOA TOTAL Controls
GSA_VA
Monitor immunoassay screening procedures with multi-constituent urine toxicology controls.
Supplier:  Thermo Scientific™ DOAT6
 View more versions of this product
Catalog No. 23-139-336
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Monitor immunoassay screening procedures with liquid, ready-to-use Thermo Scientific™ MAS™ DOA TOTAL Controls. These are multi-constituent urine toxicology controls, offering 19 analytes with four distinct levels at drug concentrations 25% below and above commonly used screening and SAMHSA cutoffs. A drug-free level and high positive level are also available (six levels total).
  • Liquid, ready-to-use
  • Human urine based matrix
  • GC/MS confirmation values available
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions