Learn More
Thermo Scientific™ MAS™ Lipid Control
Catalog No. 23100002 Shop All Thermo Scientific Products
Change view
Click to view available options
Level:
2
Catalog No. Level
23-100002 2
1 options
Catalog No. 23-100002 Supplier Thermo Scientific™ Supplier No. LIP 202
Item Discontinued This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
Bi-level, assayed control offering a complete lipid profile
Assayed ranges for precipitate and direct HDL/LDL methods
Liquid, ready-to-use control eliminates variables due to reconstitution, mixing errors, or uncertain water quality
TRUSTED_SUSTAINABILITY
Specifications
Level 2