Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ Omni·IMMUNE™ Max Controls

Catalog No. 22158064
Change view
Click to view available options
Level:
1
2
3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
22-158-064 1
22-158-065 2
22-158-066 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-158-064 Supplier Thermo Scientific™ Supplier No. OIMM101

This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.

This item is not returnable. View return policy

Provides consolidation for routine immunoassay, tumor marker and specialty immunoassay QC processing into a single product, offering three distinct levels in a barcoded load and go tube format.

Consolidate routine immunoassay, tumor marker and specialty immunoassay QC processing into a single product offering three distinct levels with Thermo Scientific MAS Omni·IMMUNE Max Controls. Value assignment is provided for key integrated instrument systems. Load-and-Go barcoded

  • Three distinct levels
  • Liquid, ready-to-use
  • Human-serum based matrix
  • Assayed values available for most analyzers
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Human Serum
Type Clinical Chemistry Control
Disposable Reusable
Level 1
Sterility Non-sterile
Form Liquid Ready-to-Use

Available in select countries only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.