Learn More
Description
Consolidate routine immunoassay, tumor marker and specialty immunoassay QC processing into a single product offering three distinct levels with Thermo Scientific MAS Omni·IMMUNE Max Controls. Value assignment is provided for key integrated instrument systems. Load-and-Go barcoded
- Three distinct levels
- liquid ready-to-use
- Human-serum based matrix
- Assayed values available for most analyzers
- Load-and-Go barcoded¹ tubes for improved efficiency and daily time savings
Required addition:
- ¹Barcodes available for selected instruments.
- Availability of products in each country depends on local regulatory marketing authorization status.
Specifications
Specifications
| Sample Type | Human Serum |
| Type | Clinical Chemistry Control |
| Disposable | Reusable |
| Level | 3 |
| Sterility | Non-sterile |
| Form | Liquid Ready-to-Use |
Available in select countries only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.