Learn More
Thermo Scientific™ MAS™ SARS-CoV-2 Serology Quality Controls
Catalog No. 10028306 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
SARS-CoV-2 IgG Positive Control
SARS-CoV-2 Negative Control
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
10-028-306 SARS-CoV-2 Negative Control
10-028-305 SARS-CoV-2 IgG Positive Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-028-306 Supplier Thermo Scientific™ Supplier No. 10028306
Add to Cart
Add to Cart
This item is not returnable. View return policy
Assess the performance of serological assays for SARS-CoV-2 infections using the Thermo Scientific MAS SARS-CoV-2 Serology Quality Controls. These third-party controls include positive controls for SARS-CoV-2 and a negative control for accuracy.
Assess the performance of serological assays for SARS-CoV-2 infections using the Thermo Scientific MAS SARS-CoV-2 Serology Quality Controls. These third-party controls include individual positive control for SARS-CoV-2 as well as a negative control for accuracy.
  • 21 days open vial stability at 2°C to 8°C
  • Ready to use straight from the refrigerator
  • Human Plasma matrix
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Type SARS-CoV-2 Negative Control
Packaging One box of control contains 5 tubes of negative control
Quantity 5 x 3.0 mL