Learn More
Thermo Scientific™ MAS™ T-Marker™ Controls
Monitor the performance of various cancer antigen tests on automated instruments with liquid assayed, multi-analyte Thermo Scientific™ MAS™ T-Marker Controls.
Supplier: Thermo Scientific™ TUM202
Description
Monitor the performance of various cancer antigen tests on automated instruments with liquid assayed, multi-analyte Thermo Scientific™ MAS™ T-Marker Controls. These controls contain more 24 analytes, including a complete offering of cancer-associated antigens.
- Liquid, ready to use; no additional preparation required
- Human serum matrix
- Includes CA 15-3, CA 19-9, CA 72-4, CA-125, CA 27-29
- Assayed values available for most major analyzers.