Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ UA Controls, 4 x 60mL
GSA_VA

Monitor urine dipstick and microscopic components with bi-level controls with improved room temperature storage capability.

Supplier:  Thermo Scientific™ UAB260

Catalog No. 23-038-385

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.

This item is not returnable. View return policy

Description

Description

Monitor urine dipstick and microscopic components with Thermo Scientific™ MAS™ UA Controls, bi-level controls with improved room temperature storage capability. Liquid and ready-to-use, this urine control is assayed for most major reagents strips and strip systems. It includes 10 commonly-assayed dipstick tests plus Microalbumin, Creatinine and hCG.

Ordering Information:
Available in convenient 15mL dropper bottles
TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.