Learn More
Thermo Scientific™ MAS™ UA Controls, Assayed, Level 1, Normal, 6 x 15mL
GSA_VA
Monitor urine dipstick and microscopic components with bi-level controls with improved room temperature storage capability.
Supplier:  Thermo Scientific™ UAB115
Catalog No. 23-038-380
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Monitor urine dipstick and microscopic components with Thermo Scientific™ MAS™ UA Controls, bi-level controls with improved room temperature storage capability. Liquid and ready-to-use, this urine control is assayed for most major reagents strips and strip systems. It includes 10 commonly-assayed dipstick tests plus Microalbumin, Creatinine and hCG.
Ordering Information:
Available in convenient 15mL dropper bottles
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions