Learn More
Aviva Systems Biology Corp MAS1 Antibody - middle region (ARP51197_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51197_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAS1. Alternate names: MAS, MGC119966ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-53652