Learn More
Medchemexpress LLC HY-10209 200mg , Masitinib AB1010 CAS:790299-79-5 Purity:98%
SDP
Catalog No. 501492575
Encompass_Preferred
Medchemexpress, HY-10209 200mg Masitinib AB1010 CAS:790299-79-5 Formula:C28H30N6OS IC50: 200 nM (Kit), 540 nM (PDGFRα), 800 nM (PDGFRβ) Purity:98% Masitinib is a novel inhibitor for Kit and PDGFRα/β with IC 50 of 200 nM and 540 nM/800 nM, and has weak inhibition to ABL and c-Fms. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2575 Supplier Medchemexpress LLC Supplier No. HY10209200MG
Medchemexpress, HY-10209 200mg Masitinib AB1010 CAS:790299-79-5 Formula:C28H30N6OS IC50: 200 nM (Kit), 540 nM (PDGFRα), 800 nM (PDGFRβ) Purity:98% Masitinib is a novel inhibitor for Kit and PDGFRα/β with IC 50 of 200 nM and 540 nM/800 nM, and has weak inhibition to ABL and c-Fms. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.