Learn More
Apexbio Technology LLC Masitinib (AB1010), 100mg. Cas: 790299-79-5 MFCD: MFCD09954132. Tyrosine kinase inhibitor, potent and selective
SDP
Supplier:  Apexbio Technology LLC A2942100
Encompass_Preferred
The stem cell factor receptor (KIT) is a therapeutic target for the cancer, mastocytosis, and inflammatory diseases treatment. Masitinib (AB1010) is a novel phenylaminothiazole-type tyrosine kinase inhibitor that targets KIT. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4765