Learn More
Selleck Chemical LLC Masitinib (AB1010) 25mg 790299-79-5
SDP
Supplier:  Selleck Chemical LLC S106425MG
Encompass_Preferred
Masitinib is a novel inhibitor for Kit (c-Kit) and PDGFR?/? with IC50 of 200 nM and 540 nM/800 nM, weak inhibition to ABL and c-Fms. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-649-5