Learn More
Medchemexpress LLC Masitinib mesylate | 1048007-93-7 | MFCD12546334 | 100.0% | 594.75 g/mol | C29H34N6O4S2 | 100 MG
SDP
Catalog No. 5000248452
Encompass_Preferred
Masitinib mesylate is the mesylate salt of masitinib, a selective, orally bioavailable tyrosine kinase inhibitor targeting c-Kit and related kinases. It is supplied as a research-grade analytical standard for preclinical and in vitro studies, with high purity and characterized physicochemical properties.
Catalog No. 50-002-48452 Supplier Medchemexpress LLC Supplier No. HY10209A100MG
Masitinib mesylate is the mesylate salt of masitinib, a selective, orally bioavailable tyrosine kinase inhibitor targeting c-Kit and related kinases. It is supplied as a research-grade analytical standard for preclinical and in vitro studies, with high purity and characterized physicochemical properties.