Learn More
Medchemexpress LLC Masitinib mesylate | 1048007-93-7 | MFCD12546334 | 100.0% | 594.75 g/mol | C29H34N6O4S2 | 25 MG
SDP
Catalog No. 5000245884
Encompass_Preferred
Masitinib mesylate is a research-grade small-molecule tyrosine kinase inhibitor primarily targeting c-Kit and used in preclinical studies to assess antiproliferative and pro-apoptotic effects. It is supplied as a high-purity solid with documented solubility and storage recommendations for research use.
  • Potent inhibitor of c-Kit with reported IC50 ≈ 200 nM.
  • High reported purity suitable for analytical and biological studies.
  • Supplied as a white to yellow solid with characterized physical properties.
  • Documented solubility in DMSO and acidic aqueous media for formulation.
  • Storage recommendations provided to preserve stability in solvent and solid form.
Catalog No. 50-002-45884 Supplier Medchemexpress LLC Supplier No. HY10209A25MG
Add to Cart
Add to Cart
Masitinib mesylate is a research-grade small-molecule tyrosine kinase inhibitor primarily targeting c-Kit and used in preclinical studies to assess antiproliferative and pro-apoptotic effects. It is supplied as a high-purity solid with documented solubility and storage recommendations for research use.
  • Potent inhibitor of c-Kit with reported IC50 ≈ 200 nM.
  • High reported purity suitable for analytical and biological studies.
  • Supplied as a white to yellow solid with characterized physical properties.
  • Documented solubility in DMSO and acidic aqueous media for formulation.
  • Storage recommendations provided to preserve stability in solvent and solid form.