Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Masitinib mesylate | 1048007-93-7 | MFCD12546334 | 100.0% | 594.75 g/mol | C29H34N6O4S2 | 5 MG
SDP

Catalog No. 5000246544
Encompass_Preferred

Masitinib mesylate is the mesylate salt form of masitinib, a small-molecule tyrosine kinase inhibitor. It is supplied as a high-purity analytical and research compound for in vitro and analytical applications and selectively inhibits c-Kit, PDGFR, and Src family kinases.

  • Purity 99.98%.
  • Molecular weight 594.75 g/mol.
  • Molecular formula C29H34N6O4S2.
  • Solubility in DMSO ≥ 30 mg/mL.
  • Intended for research and analytical use.
  • Available in a 5 mg pack size.

Catalog No. 50-002-46544 Supplier Medchemexpress LLC Supplier No. HY10209A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Masitinib mesylate is the mesylate salt form of masitinib, a small-molecule tyrosine kinase inhibitor. It is supplied as a high-purity analytical and research compound for in vitro and analytical applications and selectively inhibits c-Kit, PDGFR, and Src family kinases.

  • Purity 99.98%.
  • Molecular weight 594.75 g/mol.
  • Molecular formula C29H34N6O4S2.
  • Solubility in DMSO ≥ 30 mg/mL.
  • Intended for research and analytical use.
  • Available in a 5 mg pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.