Learn More
Medchemexpress LLC HY-10209A 200mg Medchemexpress, Masitinib (mesylate) AB-1010 mesylate CAS:1048007-93-7 Purity:98%
SDP
Catalog No. 501637084
Encompass_Preferred
Medchemexpress,HY-10209A 200mg Masitinib (mesylate) AB-1010 mesylate CAS:1048007-93-7 Formula:C29H34N6O4S2 Masitinib mesylate is a novel inhibitor for Kit and PDGFRα/β with IC50 of 200 nM and 540 nM/800 nM, and has weak inhibition to ABL and c-Fms. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7084 Supplier Medchemexpress LLC Supplier No. HY10209A200MG
Medchemexpress,HY-10209A 200mg Masitinib (mesylate) AB-1010 mesylate CAS:1048007-93-7 Formula:C29H34N6O4S2 Masitinib mesylate is a novel inhibitor for Kit and PDGFRα/β with IC50 of 200 nM and 540 nM/800 nM, and has weak inhibition to ABL and c-Fms. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.