Learn More
Dentec Safety™ Mask Cover Window
SDP
Catalog No. 19222487 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
25/package
Catalog No.
19-222-487 25/package
1 options
Catalog No. 19-222-487 Supplier Dentec Safety™ Supplier No. 1307096
Add to Cart
Add to Cart
Mask cover window protects primary lenses from splash and scratches.
  • For 130M Series full face respirator
  • Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator facepiece
Specifications
Material Mylar
Product Type Mask Cover Window
For Use With (Equipment) 130M Series full face respirator
Includes Helmets, Attachments
Model 1307096
Quantity 25/Pk.