Learn More
Epredia™ Mask Cytoslides
Catalog No. B5991050 Shop All Epredia Products
Change view
Click to view available options
Description:
Double circle
Single circle
Color:
Black
Blue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Color
B5991050 Double circle Blue
59-910-57 Single circle Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B5991050 Supplier Epredia™ Supplier No. B5991050
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for immunofluorescence. Masking blocks out UV light to enhance microscopic viewing.
  • For immunofluorescence
  • Blocks out UV light
Specifications
No. per Pack 100
Well Count 2
Color Blue
Description Double circle
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov