Learn More
Med Vet International Procedure Mask Pleated Earloops One Size Fits Most Kid Design (Happy Face Print) Nonsterile Not Rated Pediatric, 15150, 750/Case
Catalog No. 502831513
Encompass_Preferred
Child-sized to improve fit for children High filtration for improved protection Soft comfortable earloops provide easier donning Designed specifically to be used by children ages 3 - 12
Catalog No. 50-283-1513 Supplier Med Vet International Supplier No. 706259CS
Child-sized to improve fit for children High filtration for improved protection Soft comfortable earloops provide easier donning Designed specifically to be used by children ages 3 - 12