Learn More
Med Vet International Particulate Respirator Mask 3M industrial N95 Cup Elastic Strap One Size Fits Most White Nonsterile Not Rated Adult, 8210, 160/Case
Catalog No. 502831942
Encompass_Preferred
The 3M™ Particulate Respirator N95 filters out at least 95% of non-oil based particles, keeping air clean and breathable Dual straps attached at two welded points keeps the mask securely in place for a reliable seal Foam cushioning around the nose offers additional comfort This 3M™ particulate mask is lightweight for easier acceptance, even when worn for prolonged periods Electrostatic media allows for easy air flow and comfortable breathing
Catalog No. 50-283-1942 Supplier Med Vet International Supplier No. 794716CS
The 3M™ Particulate Respirator N95 filters out at least 95% of non-oil based particles, keeping air clean and breathable Dual straps attached at two welded points keeps the mask securely in place for a reliable seal Foam cushioning around the nose offers additional comfort This 3M™ particulate mask is lightweight for easier acceptance, even when worn for prolonged periods Electrostatic media allows for easy air flow and comfortable breathing