Learn More
Med Vet International Progear N95 Particulate Filter Respirator And Surgical Mask, Astm Level 3, White, Small, Rp88010, 50/Box
Catalog No. 502830733
Encompass_Preferred
Particulate Respirator / Surgical Mask ProGear Medical N95 Flat Fold Pouch Elastic Strap Small White NonSterile ASTM Level 3 Adult Does not contain cellulose Filters out 99% of particles down to 0.1 microns Snugly conforms to many different face-shapes Large filter area allows for easy breathing and speaking
Catalog No. 50-283-0733 Supplier Med Vet International Supplier No. 1181773BX
Particulate Respirator / Surgical Mask ProGear Medical N95 Flat Fold Pouch Elastic Strap Small White NonSterile ASTM Level 3 Adult Does not contain cellulose Filters out 99% of particles down to 0.1 microns Snugly conforms to many different face-shapes Large filter area allows for easy breathing and speaking