Learn More
Med Vet International 3M 6000 Reusable Respirator industrial Half Face 4 Point Adjustable Head Strap Medium Gray / White, 7000001933, 24/Case
Catalog No. 502829895
Encompass_Preferred
This product should not be used in a clinical setting where the risk of infection through inhalation exposure is high This half facepiece reusable respirator offers reliable and convenient respiratory protection and is suitable for many situations, helping provide protection against particulates and a wide variety of gases and vapors according to NIOSH approvals Comfortable soft, lightweight facepiece with easy-to-adjust head straps NIOSH approved for negative pressure air purifying and supplied air dual airline applications Can be disassembled, cleaned and reused Replaceable cartridges and filters allow reuse of facepiece for cost savings
Catalog No. 50-282-9895 Supplier Med Vet International Supplier No. 1088071CS
This product should not be used in a clinical setting where the risk of infection through inhalation exposure is high This half facepiece reusable respirator offers reliable and convenient respiratory protection and is suitable for many situations, helping provide protection against particulates and a wide variety of gases and vapors according to NIOSH approvals Comfortable soft, lightweight facepiece with easy-to-adjust head straps NIOSH approved for negative pressure air purifying and supplied air dual airline applications Can be disassembled, cleaned and reused Replaceable cartridges and filters allow reuse of facepiece for cost savings